Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129305.3 | NP_001122777.1 | 9 - 10 | 1219 | 95 | |
NM_001307091.3 | NP_001294020.1 | 12 - 13 | 1469 | 95 | ||
NM_001307092.4 | NP_001294021.1 | 11 - 12 | 1340 | 95 | ||
NM_001380456.1 | NP_001367187.1 | 10 - 11 | 1354 | 95 | ||
NM_001380457.2 | NP_001367146.1 | 11 - 12 | 1408 | 95 | ||
NM_171418.7 | NP_741488.1 | 11 - 12 | 1389 | 95 | ||
NM_171918.7 | NP_741489.1 | 11 - 12 | 1346 | 95 | ||
NM_171919.7 | NP_741490.1 | 11 - 12 | 1385 | 95 | ||
GenBank mRNA | X68070.1 | - | 1398 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |