Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sru-26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276870.1 | NP_001263799.1 | 2 - 3 | 454 | 102 | |
NM_171421.3 | NP_741493.2 | 2 - 3 | 539 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |