Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028998.4 | NP_001024169.1 | 5 - 6 | 827 | 73 | |
NM_001028999.5 | NP_001024170.1 | 5 - 6 | 1178 | 73 | ||
NM_001029002.4 | NP_001024173.1 | 4 - 5 | 712 | 73 | ||
NM_001029004.5 | NP_001024175.1 | 4 - 5 | 601 | 73 | ||
NM_001313278.3 | NP_001300207.1 | 4 | 601 | 73 | ||
NM_001380634.1 | NP_001367924.1 | 5 - 6 | 827 | 73 | ||
NM_001380635.1 | NP_001367923.1 | 5 - 6 | 829 | 73 | ||
NM_001383298.1 | 5 - 6 | 821 | 73 | |||
GenBank mRNA | AF427474.1 | - | 5 - 6 | 849 | 73 | |
AF427475.1 | - | 5 - 6 | 849 | 73 | ||
AF427476.1 | - | 5 - 6 | 1032 | 73 | ||
AF427477.1 | - | 5 - 6 | 1032 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |