Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pst-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028889.4 | NP_001024060.1 | 2 - 3 | 203 | 112 | |
NM_001373028.2 | NP_001360551.1 | 3 | 180 | 112 | ||
NM_001383312.1 | 2 - 3 | 175 | 112 | |||
NM_171463.5 | NP_741545.2 | 2 - 3 | 173 | 112 | ||
NM_171464.5 | NP_741546.3 | 2 - 3 | 535 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |