Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029029.5 | NP_001024200.1 | 3 - 4 | 636 | 90 | |
NM_001359765.3 | NP_001346723.1 | 537 | 90 | |||
NM_001359766.3 | NP_001346716.1 | 537 | 90 | |||
NM_001359767.3 | NP_001346722.1 | 537 | 90 | |||
NM_001359768.3 | NP_001346715.1 | 537 | 90 | |||
NM_001359777.3 | NP_001346724.1 | 3 - 4 | 624 | 90 | ||
NM_001373034.1 | NP_001360072.1 | 630 | 90 | |||
NM_001380656.1 | NP_001367914.1 | 537 | 90 | |||
NM_001380657.1 | NP_001367912.1 | 630 | 90 | |||
NM_001380658.2 | NP_001367913.1 | 537 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |