Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gon-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171475.5 | NP_741557.2 | 13 - 14 | 2669 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |