Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: xpo-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171484.7 | NP_741567.1 | 3 - 4 | 841 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |