Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sth-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171489.7 | NP_741574.1 | 3 - 4 | 240 | 112 | |
GenBank mRNA | AB072927.1 | - | 3 - 4 | 204 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |