Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atg-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171491.5 | NP_741576.1 | 2 - 3 | 161 | 93 | |
NM_171492.6 | NP_741577.1 | 2 - 3 | 163 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |