Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atg-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171491.5 | NP_741576.1 | 5 - 6 | 987 | 84 | |
NM_171492.6 | NP_741577.1 | 5 - 6 | 989 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |