Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mod-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028863.5 | NP_001024034.1 | 1 - 2 | 108 | 82 | |
NM_001028864.4 | NP_001024035.1 | 1 - 2 | 60 | 82 | ||
NM_171495.6 | NP_741580.1 | 1 - 2 | 101 | 82 | ||
GenBank mRNA | AF303088.1 | - | 1 - 2 | 60 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |