Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mod-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028863.5 | NP_001024034.1 | 5 - 6 | 951 | 131 | |
NM_001028864.4 | NP_001024035.1 | 5 - 6 | 903 | 131 | ||
NM_171495.6 | NP_741580.1 | 5 - 6 | 944 | 131 | ||
GenBank mRNA | AF303088.1 | - | 5 - 6 | 903 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |