Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spas-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269186.3 | NP_001256115.1 | 5 - 6 | 704 | 90 | |
NM_171501.6 | NP_741586.1 | 4 - 5 | 518 | 90 | ||
NR_136364.1 | 3 - 4 | 883 | 90 | |||
GenBank mRNA | AB287436.1 | - | 5 - 6 | 690 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |