Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028662.4 | NP_001023833.1 | 4 - 5 | 410 | 79 | |
NM_001028663.6 | NP_001023834.1 | 7 - 8 | 1188 | 79 | ||
NM_001047640.4 | NP_001041105.1 | 5 - 6 | 952 | 79 | ||
NM_171533.6 | NP_741621.1 | 7 - 8 | 1188 | 79 | ||
GenBank mRNA | AF178536.1 | - | 7 - 8 | 1190 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Caenorhabditis elegans Probe spans exons |