Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sma-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136431.4 | NP_001129903.1 | 16 - 17 | 12142 | 123 | |
NM_001269453.3 | NP_001256382.1 | 14 - 15 | 11893 | 123 | ||
NM_001269454.2 | NP_001256383.1 | 15 - 16 | 12054 | 123 | ||
NM_001373124.2 | NP_001360523.1 | 13 | 12121 | 123 | ||
NM_001383415.1 | 15 - 16 | 12060 | 123 | |||
NM_171544.7 | NP_741632.2 | 17 - 18 | 12566 | 123 | ||
GenBank mRNA | AF053496.1 | - | 16 - 17 | 12142 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |