Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hot-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171552.4 | NP_741640.1 | 1 - 2 | 226 | 117 | |
GenBank mRNA | AF324053.1 | - | 1 - 2 | 220 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |