Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slo-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029088.2 | NP_001024259.1 | 17 - 18 | 3047 | 82 | |
NM_001029089.3 | NP_001024260.1 | 16 - 17 | 2987 | 82 | ||
NM_001029090.5 | NP_001024261.1 | 17 - 18 | 3026 | 82 | ||
NM_001269872.3 | NP_001256801.1 | 18 - 19 | 3026 | 82 | ||
NM_001269873.3 | NP_001256802.1 | 18 - 19 | 3068 | 82 | ||
NM_001269874.3 | NP_001256803.1 | 17 - 18 | 3002 | 82 | ||
NM_001269875.4 | NP_001256804.1 | 18 - 19 | 3092 | 82 | ||
NM_001269876.3 | NP_001256805.1 | 17 - 18 | 3008 | 82 | ||
NM_001269877.3 | NP_001256806.1 | 16 - 17 | 2942 | 82 | ||
NM_001269878.3 | NP_001256807.1 | 18 - 19 | 3068 | 82 | ||
NM_001269879.4 | NP_001256808.1 | 17 - 18 | 3047 | 82 | ||
NM_001269880.3 | NP_001256809.1 | 18 - 19 | 3047 | 82 | ||
NM_001269881.3 | NP_001256810.1 | 17 - 18 | 2981 | 82 | ||
GenBank mRNA | AF431891.1 | - | 17 - 18 | 3008 | 82 | |
AF431892.1 | - | 16 - 17 | 2942 | 82 | ||
AF431893.1 | - | 17 - 18 | 2981 | 82 | ||
JQ582463.1 | - | 18 - 19 | 3068 | 82 | ||
JQ606809.1 | - | 18 - 19 | 3047 | 82 | ||
JQ606810.1 | - | 16 - 17 | 2942 | 82 | ||
JQ606812.1 | - | 17 - 18 | 2981 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |