Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slo-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029090.5 | NP_001024261.1 | 9 - 10 | 1798 | 73 | |
NM_001269873.3 | NP_001256802.1 | 9 - 10 | 1753 | 73 | ||
NM_001269874.3 | NP_001256803.1 | 9 - 10 | 1753 | 73 | ||
NM_001269875.4 | NP_001256804.1 | 9 - 10 | 1798 | 73 | ||
NM_001269876.3 | NP_001256805.1 | 9 - 10 | 1753 | 73 | ||
NM_001269877.3 | NP_001256806.1 | 9 - 10 | 1753 | 73 | ||
GenBank mRNA | AF431893.1 | - | 9 - 10 | 1753 | 73 | |
JQ582463.1 | - | 9 - 10 | 1753 | 73 | ||
JQ606809.1 | - | 9 - 10 | 1753 | 73 | ||
JQ606810.1 | - | 9 - 10 | 1753 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |