Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029058.4 | NP_001024229.1 | 3 - 4 | 308 | 74 | |
NM_001029059.5 | NP_001024230.1 | 3 - 4 | 305 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |