Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bed-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029056.5 | NP_001024227.1 | 5 - 6 | 695 | 78 | |
NM_001029057.2 | NP_001024228.1 | 5 - 6 | 694 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |