Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tsp-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171602.3 | NP_741692.1 | 1 - 2 | 145 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |