Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncam-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171615.7 | NP_741707.3 | 6 - 7 | 1873 | 68 | |
NM_171616.5 | NP_741708.1 | 6 - 7 | 757 | 68 | ||
NM_171617.5 | NP_741709.1 | 5 - 6 | 897 | 68 | ||
GenBank mRNA | AF456360.2 | - | 6 - 7 | 781 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |