Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cgef-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171641.6 | NP_741736.2 | 13 - 14 | 1448 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |