Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klp-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373256.3 | NP_001359863.1 | 13 | 2652 | 114 | |
NM_001373257.3 | NP_001359864.1 | 12 - 13 | 2607 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Amplicon spans exons and probe does not span exons |