Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbc-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373289.3 | NP_001359912.1 | 16 - 17 | 1775 | 123 | |
NM_001373290.4 | NP_001360675.1 | 13 - 14 | 1486 | 123 | ||
NM_171949.8 | NP_741779.1 | 14 - 15 | 1649 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |