Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ags-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322618.4 | NP_001309689.1 | 2 - 3 | 178 | 73 | |
NM_001322619.4 | NP_001309690.1 | 1 - 2 | 49 | 73 | ||
NM_001383577.1 | 8 - 9 | 1064 | 73 | |||
NM_171684.6 | NP_741787.1 | 8 - 9 | 988 | 73 | ||
NR_136376.1 | 8 - 9 | 1048 | 73 | |||
NR_136377.1 | 6 - 7 | 894 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |