Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chtl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380966.2 | NP_001367416.1 | 8 - 9 | 2150 | 106 | |
NM_001380967.2 | NP_001367417.1 | 8 - 9 | 2158 | 106 | ||
NM_171686.7 | NP_741790.2 | 10 - 11 | 2052 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |