Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mec-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001368504.4 | NP_001355460.1 | 4 | 877 | 80 | |
NM_001368505.3 | NP_001355457.1 | 8 - 9 | 946 | 80 | ||
NM_001368507.2 | NP_001355459.1 | 8 - 9 | 946 | 80 | ||
NM_001368511.3 | NP_001355478.1 | 8 - 9 | 922 | 80 | ||
NM_001368515.4 | NP_001355479.1 | 8 | 898 | 80 | ||
NM_001380971.2 | NP_001368710.1 | 8 - 9 | 1063 | 80 | ||
NM_001380972.1 | NP_001368712.1 | 9 - 10 | 1123 | 80 | ||
NM_001380973.1 | NP_001368715.1 | 9 - 10 | 1123 | 80 | ||
NM_001380974.1 | NP_001368713.1 | 8 - 9 | 922 | 80 | ||
NM_001380975.1 | NP_001368716.1 | 8 - 9 | 922 | 80 | ||
NM_001380976.1 | NP_001368714.1 | 8 - 9 | 898 | 80 | ||
NM_001380977.1 | NP_001368717.1 | 8 - 9 | 898 | 80 | ||
NM_171691.6 | NP_741797.1 | 9 - 10 | 1166 | 80 | ||
GenBank mRNA | U26735.1 | - | 9 - 10 | 1160 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |