Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pak-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029206.3 | NP_001024377.1 | 8 - 9 | 1378 | 68 | |
NM_001029207.4 | NP_001024378.1 | 8 - 9 | 1367 | 68 | ||
NM_001029208.5 | NP_001024379.1 | 7 - 8 | 1221 | 68 | ||
GenBank mRNA | D83215.1 | - | 8 - 9 | 1364 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |