Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gar-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029230.3 | NP_001024401.1 | 12 - 13 | 1365 | 63 | |
NM_001029231.3 | NP_001024402.1 | 13 - 14 | 1528 | 63 | ||
NM_001029232.2 | NP_001024403.1 | 12 - 13 | 1365 | 63 | ||
NM_001373329.1 | NP_001360660.1 | 12 - 13 | 1365 | 63 | ||
NM_001373330.1 | NP_001360659.1 | 13 - 14 | 1458 | 63 | ||
GenBank mRNA | AF075245.1 | - | 12 - 13 | 1365 | 63 | |
AF117300.1 | - | 13 - 14 | 1458 | 63 | ||
AF117301.1 | - | 12 - 13 | 1365 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Caenorhabditis elegans Probe spans exons |