Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kqt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171709.6 | NP_741819.1 | 9 - 10 | 989 | 79 | |
NM_171710.7 | NP_741820.1 | 8 - 9 | 1004 | 79 | ||
GenBank mRNA | AY572974.1 | - | 8 - 9 | 835 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |