Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhx-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029391.4 | NP_001024562.1 | 6 - 7 | 1259 | 74 | |
NM_001381015.1 | NP_001367515.1 | 6 - 7 | 1263 | 74 | ||
GenBank mRNA | AF497826.1 | - | 6 - 7 | 1281 | 74 | |
AF497827.1 | - | 3 - 4 | 1001 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |