Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: inx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029236.3 | NP_001024407.1 | 5 - 6 | 598 | 117 | |
NM_171716.5 | NP_741826.1 | 5 - 6 | 598 | 117 | ||
NR_131721.1 | 5 - 6 | 542 | 117 | |||
NR_131722.1 | 5 - 6 | 542 | 117 | |||
NR_131723.1 | 4 - 5 | 522 | 117 | |||
NR_131724.1 | 4 - 5 | 522 | 117 | |||
GenBank mRNA | U59212.1 | - | 5 - 6 | 598 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |