Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029291.3 | NP_001024462.1 | 8 - 9 | 1101 | 120 | |
NM_001029292.3 | NP_001024463.1 | 8 - 9 | 1080 | 120 | ||
NM_001029293.3 | NP_001024464.1 | 8 - 9 | 1272 | 120 | ||
NM_001029294.4 | NP_001024465.1 | 8 - 9 | 1295 | 120 | ||
NM_001029295.5 | NP_001024466.1 | 8 - 9 | 1272 | 120 | ||
NM_001029296.8 | NP_001024467.1 | 8 - 9 | 1156 | 120 | ||
NM_001373367.3 | NP_001360693.1 | 7 | 1080 | 120 | ||
GenBank mRNA | AF083643.1 | - | 8 - 9 | 1080 | 120 | |
AF083644.1 | - | 8 - 9 | 1080 | 120 | ||
AF083647.1 | - | 8 - 9 | 1272 | 120 | ||
AF083648.1 | - | 8 - 9 | 1272 | 120 | ||
AF083651.1 | - | 8 - 9 | 1272 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |