Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnt-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373379.1 | NP_001360658.1 | 5 - 6 | 508 | 75 | |
NM_001373380.2 | NP_001359846.1 | 5 - 6 | 460 | 75 | ||
NM_001373381.2 | NP_001359848.1 | 4 - 5 | 3135 | 75 | ||
NM_001373382.2 | NP_001360657.1 | 4 - 5 | 3134 | 75 | ||
NM_001373383.1 | NP_001359847.1 | 4 - 5 | 3135 | 75 | ||
NM_001383610.1 | 4 - 5 | 379 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |