Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171742.5 | NP_741860.2 | 1 - 2 | 141 | 73 | |
NM_171743.6 | NP_741861.2 | 1 - 2 | 143 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |