Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pkc-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029345.6 | NP_001024516.1 | 6 - 7 | 722 | 111 | |
NM_001029346.4 | NP_001024517.1 | 6 - 7 | 723 | 111 | ||
NM_001129650.5 | NP_001123122.1 | 8 - 9 | 1067 | 111 | ||
NM_001373417.3 | NP_001360728.1 | 8 | 829 | 111 | ||
NM_001373418.2 | NP_001360727.1 | 7 - 8 | 1060 | 111 | ||
NM_001373419.3 | NP_001359883.1 | 7 - 8 | 907 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |