Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171756.4 | NP_741878.1 | 5 - 6 | 514 | 88 | |
NM_171757.7 | NP_741879.1 | 5 - 6 | 733 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |