Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373456.2 | NP_001360793.1 | 13 - 14 | 1738 | 96 | |
NM_001373457.2 | NP_001360791.1 | 11 - 12 | 1618 | 96 | ||
NM_001373458.3 | NP_001360790.1 | 12 | 1755 | 96 | ||
NM_001373459.2 | NP_001360794.1 | 10 - 11 | 1565 | 96 | ||
NM_001373460.3 | NP_001360792.1 | 11 - 12 | 1475 | 96 | ||
NM_171758.5 | NP_741880.1 | 11 - 12 | 1599 | 96 | ||
NM_171759.6 | NP_741881.1 | 10 - 11 | 1663 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |