Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nucb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171763.5 | NP_741885.1 | 4 - 5 | 421 | 74 | |
NM_171970.7 | NP_741886.1 | 4 - 5 | 421 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |