Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhx-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029549.4 | NP_001024720.1 | 5 - 6 | 794 | 117 | |
NM_001373485.2 | NP_001359642.1 | 5 - 6 | 794 | 117 | ||
GenBank mRNA | AF497828.1 | - | 5 - 6 | 815 | 117 | |
AF497829.1 | - | 5 - 6 | 815 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |