Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ifa-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083295.4 | NP_001076764.1 | 4 - 5 | 444 | 72 | |
NM_001270210.4 | NP_001257139.1 | 5 - 6 | 517 | 72 | ||
NM_001373503.1 | NP_001360640.1 | 5 - 6 | 553 | 72 | ||
NM_171777.8 | NP_741902.1 | 5 - 6 | 530 | 72 | ||
NM_171778.7 | NP_741903.1 | 5 - 6 | 572 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |