Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sma-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047832.3 | NP_001041297.1 | 3 - 4 | 387 | 89 | |
NM_001381114.2 | NP_001366649.1 | 3 - 4 | 932 | 89 | ||
NM_001383653.1 | 3 - 4 | 414 | 89 | |||
NM_171782.6 | NP_741908.1 | 2 - 3 | 362 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |