Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373531.3 | NP_001360646.1 | 4 | 1379 | 113 | |
NM_001373534.4 | NP_001360782.1 | 4 | 1447 | 113 | ||
NM_001373535.3 | NP_001360784.1 | 3 - 4 | 1082 | 113 | ||
NM_001381120.2 | NP_001368763.1 | 2 - 3 | 981 | 113 | ||
NM_001381121.2 | NP_001368761.1 | 2 - 3 | 977 | 113 | ||
NM_001381122.2 | NP_001367017.1 | 2 - 3 | 977 | 113 | ||
NM_001381123.2 | NP_001368762.1 | 3 - 4 | 1351 | 113 | ||
NM_171974.7 | NP_741912.1 | 4 - 5 | 1430 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |