Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: obr-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383668.1 | 6 - 7 | 758 | 68 | ||
NM_171794.6 | NP_741923.1 | 5 - 6 | 619 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |