Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: obr-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383668.1 | 10 - 11 | 2055 | 115 | ||
NM_171794.6 | NP_741923.1 | 9 - 10 | 1916 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |