Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eor-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029299.6 | NP_001024470.1 | 10 - 11 | 1859 | 82 | |
NM_001029300.6 | NP_001024471.1 | 9 - 10 | 1814 | 82 | ||
NM_001313378.4 | NP_001300307.1 | 9 | 1784 | 82 | ||
GenBank mRNA | AF502570.1 | - | 10 - 11 | 1806 | 82 | |
AF519109.1 | - | 10 - 11 | 1784 | 82 | ||
AF519110.1 | - | 9 - 10 | 1739 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |