Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356849.3 | NP_001343766.1 | 5 | 509 | 66 | |
NM_001381442.1 | NP_001368664.1 | 2 - 3 | 338 | 66 | ||
NM_001383853.1 | 4 - 5 | 473 | 66 | |||
NM_062785.5 | NP_495186.3 | 5 - 6 | 537 | 66 | ||
NM_171850.7 | NP_741007.2 | 5 - 6 | 556 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |