Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cor-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171892.7 | NP_741269.1 | 8 - 9 | 1680 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Caenorhabditis elegans Probe spans exons |