Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cor-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171893.6 | NP_741270.1 | 7 - 8 | 1490 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Caenorhabditis elegans Probe spans exons |